siRNA Details
| Gene Symbol: A930024O17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020888.1 | - | 1 | 546 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU502958.1 | - | 1 | 92 | Chr. 11 - 117385046 - 117385393 [+] on Build GRCm38 | |

A930024O17Rik
Genomic Map