siRNA Details
| Gene Symbol: Ccdc30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005616.1 | - | 1 | 508 | ||
| AK079015.1 | - | 1 | 519 | |||
| AK225018.1 | - | 1 | 509 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc30| Gene Symbol: Ccdc30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005616.1 | - | 1 | 508 | ||
| AK079015.1 | - | 1 | 519 | |||
| AK225018.1 | - | 1 | 509 | |||