siRNA Details
Gene Symbol: 2010007H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008147.1 | - | 1 | 432 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY708113.1 | - | 1 | 439 | Chr. 9 - 51280290 - 51280934 [+] on Build GRCm38 |
