siRNA Details
| Gene Symbol: 2010007H06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008147.1 | - | 1 | 385 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY708113.1 | - | 1 | 392 | Chr. 9 - 51280290 - 51280934 [+] on Build GRCm38 | |

2010007H06Rik
Genomic Map