siRNA Details
| Gene Symbol: 4933416E03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040498.1 | 6 | 611 | |||
| GenBank mRNA | AK016829.1 | - | 6 | 613 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716503.1 | - | 6 | 613 | Chr. 2 - 159947341 - 159981092 [+] on Build GRCm38 | |

4933416E03Rik
Genomic Map