siRNA Details
| Gene Symbol: 4933416E03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040498.1 | 7 | 965 | |||
| GenBank mRNA | AK016829.1 | - | 7 | 967 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933416E03Rik| Gene Symbol: 4933416E03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040498.1 | 7 | 965 | |||
| GenBank mRNA | AK016829.1 | - | 7 | 967 | ||