siRNA Details
| Gene Symbol: 4833412C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045954.1 | 3 | 384 | |||
| GenBank mRNA | AK014683.1 | - | 3 | 385 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4833412C05Rik| Gene Symbol: 4833412C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045954.1 | 3 | 384 | |||
| GenBank mRNA | AK014683.1 | - | 3 | 385 | ||