siRNA Details
| Gene Symbol: 4833412C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045954.1 | 1 | 72 | |||
| GenBank mRNA | AK014683.1 | - | 1 | 73 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714180.1 | - | 1 | 62 | Chr. 7 - 67784775 - 67803486 [-] on Build GRCm38 | |

4833412C05Rik
Genomic Map