siRNA Details
Gene Symbol: 4833412C15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014684.1 | - | 1 | 1437 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ238017.1 | - | 1 | 40 | Chr. 15 - 77113733 - 77114405 [-] on Build GRCm38 |
