siRNA Details
| Gene Symbol: Gm32568 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK089350.1 | - | 3 | 1087 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY764325.1 | - | 1 | 372 | Chr. 8 - 66496254 - 66496946 [+] on Build GRCm38 | |

Gm32568
Genomic Map