siRNA Details
| Gene Symbol: Gm12718 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040673.1 | 4 | 292 | |||
| GenBank mRNA | AK144477.1 | - | 4 | 293 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY733749.1 | - | 4 | 293 | Chr. 4 - 103382500 - 103492040 [+] on Build GRCm38 | |

Gm12718
Genomic Map