siRNA Details
| Gene Symbol: 4930438E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045961.1 | 6 | 652 | |||
| GenBank mRNA | AK015336.1 | - | 6 | 653 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930438E09Rik| Gene Symbol: 4930438E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045961.1 | 6 | 652 | |||
| GenBank mRNA | AK015336.1 | - | 6 | 653 | ||