siRNA Details
| Gene Symbol: 4930438E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045961.1 | 5, 6 | 368 | |||
| GenBank mRNA | AK015336.1 | - | 5, 6 | 369 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714888.1 | - | 5, 6 | 367 | Chr. 14 - 67332046 - 67397741 [-] on Build GRCm38 | |

4930438E09Rik
Genomic Map