siRNA Details
| Gene Symbol: 4930459C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110448.1 | 3 | 929 | |||
| GenBank mRNA | AK015487.1 | - | 3 | 931 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930459C07Rik| Gene Symbol: 4930459C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110448.1 | 3 | 929 | |||
| GenBank mRNA | AK015487.1 | - | 3 | 931 | ||