siRNA Details
| Gene Symbol: 4930459C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110448.1 | 3 | 546 | |||
| GenBank mRNA | AK015487.1 | - | 3 | 548 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715035.1 | - | 3 | 548 | Chr. 10 - 96226207 - 96248350 [+] on Build GRCm38 | |

4930459C07Rik
Genomic Map