siRNA Details
| Gene Symbol: 2900046F13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013655.1 | - | 1 | 674 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DV045881.1 | - | 90 | Chr. - - [-] on Build | ||

2900046F13Rik
Genomic Map