siRNA Details
| Gene Symbol: Wdr66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015152.1 | - | 1 | 506 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr66| Gene Symbol: Wdr66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015152.1 | - | 1 | 506 | ||