siRNA Details
| Gene Symbol: 4930558G05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045441.1 | 1, 2 | 103 | |||
| GenBank mRNA | AK016174.1 | - | 1, 2 | 106 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715778.1 | - | 1, 2 | 106 | Chr. X - 129051524 - 129067146 [+] on Build GRCm38 | |

4930558G05Rik
Genomic Map