siRNA Details
| Gene Symbol: 4930558K02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016179.1 | - | 6 | 522 | ||
| CF105154.1 | - | 6 | 486 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930558K02Rik| Gene Symbol: 4930558K02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016179.1 | - | 6 | 522 | ||
| CF105154.1 | - | 6 | 486 | |||