siRNA Details
| Gene Symbol: 5830400J07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017886.1 | - | 1 | 1162 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX517703.1 | - | 1 | 444 | Chr. 12 - 80865546 - 80866054 [+] on Build GRCm38 | |

5830400J07Rik
Genomic Map