siRNA Details
| Gene Symbol: 4930577N17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073429.1 | 1 | 1180 | |||
| GenBank mRNA | AK016297.1 | - | 1 | 1180 | ||
| AK017883.1 | - | 1 | 1049 | |||
| AK086057.1 | - | 1 | 1125 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB521487.2 | - | 1 | 256 | Chr. 3 - 51277684 - 51278383 [+] on Build GRCm38 | |

4930577N17Rik
Genomic Map