siRNA Details
| Gene Symbol: 4930577N17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073429.1 | 1 | 1490 | |||
| GenBank mRNA | AK016297.1 | - | 1 | 1490 | ||
| AK017883.1 | - | 1 | 1359 | |||
| AK086057.1 | - | 1 | 1435 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930577N17Rik| Gene Symbol: 4930577N17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073429.1 | 1 | 1490 | |||
| GenBank mRNA | AK016297.1 | - | 1 | 1490 | ||
| AK017883.1 | - | 1 | 1359 | |||
| AK086057.1 | - | 1 | 1435 | |||