siRNA Details
| Gene Symbol: 1810021B22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040417.1 | 4 | 611 | |||
| GenBank mRNA | AK007571.1 | - | 4 | 612 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707618.1 | - | 4 | 606 | Chr. 15 - 89071196 - 89075852 [-] on Build GRCm38 | |

1810021B22Rik
Genomic Map