siRNA Details
Gene Symbol: E530001K10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK140370.1 | - | 2 | 333 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE864434.1 | - | 2 | 242 | Chr. 2 - 94252127 - 94264649 [-] on Build GRCm38 |
