siRNA Details
| Gene Symbol: E530001K10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK140370.1 | - | 2 | 485 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE864434.1 | - | 2 | 394 | Chr. 2 - 94252127 - 94264649 [-] on Build GRCm38 | |

E530001K10Rik
Genomic Map