siRNA Details
| Gene Symbol: Gm13219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035738.1 | - | 4 | 338 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB628373.1 | - | 4 | 338 | Chr. 2 - 9352490 - 9445313 [+] on Build GRCm38 | |

Gm13219
Genomic Map