siRNA Details
| Gene Symbol: Gm32401 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080039.1 | - | 249 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB637077.1 | - | 3 | 249 | Chr. 13 - 41487130 - 41543932 [+] on Build GRCm38 | |

Gm32401
Genomic Map