siRNA Details
| Gene Symbol: Gm12298 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033539.1 | 4 | 766 | |||
| GenBank mRNA | AK131836.1 | - | 4 | 767 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12298| Gene Symbol: Gm12298 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033539.1 | 4 | 766 | |||
| GenBank mRNA | AK131836.1 | - | 4 | 767 | ||