siRNA Details
| Gene Symbol: Gm12298 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033539.1 | 3 | 464 | |||
| GenBank mRNA | AK131836.1 | - | 3 | 465 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707708.1 | - | 3 | 465 | Chr. 11 - 66905721 - 66947086 [-] on Build GRCm38 | |

Gm12298
Genomic Map