siRNA Details
| Gene Symbol: 4933436I20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017086.1 | - | 7 | 1154 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933436I20Rik| Gene Symbol: 4933436I20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017086.1 | - | 7 | 1154 | ||