siRNA Details
| Gene Symbol: 4933436H12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046208.1 | 2 | 1003 | |||
| GenBank mRNA | AK017082.1 | - | 2 | 1005 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933436H12Rik| Gene Symbol: 4933436H12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046208.1 | 2 | 1003 | |||
| GenBank mRNA | AK017082.1 | - | 2 | 1005 | ||