siRNA Details
| Gene Symbol: 4933436H12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046208.1 | 2 | 435 | |||
| GenBank mRNA | AK017082.1 | - | 2 | 437 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716760.1 | - | 2 | 437 | Chr. 7 - 68417424 - 68425180 [+] on Build GRCm38 | |

4933436H12Rik
Genomic Map