siRNA Details
| Gene Symbol: 2210011G09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008706.1 | - | 1 | 129 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY708595.1 | - | 1 | 127 | Chr. 2 - 80633740 - 80634440 [+] on Build GRCm38 | |

2210011G09Rik
Genomic Map