siRNA Details
| Gene Symbol: Gm6209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK138462.1 | - | 3 | 254 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ057533.1 | - | 3 | 254 | Chr. 3 - 50517255 - 50607222 [-] on Build GRCm38 | |

Gm6209
Genomic Map