siRNA Details
| Gene Symbol: Gm20337 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045057.1 | 3 | 927 | |||
| GenBank mRNA | AK145430.1 | - | 3 | 926 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY756110.1 | - | 3 | 489 | Chr. 12 - 80942673 - 80944916 [-] on Build GRCm38 | |

Gm20337
Genomic Map