siRNA Details
| Gene Symbol: Gm20337 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045057.1 | 1 | 97 | |||
| GenBank mRNA | AK145430.1 | - | 1 | 96 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ963819.1 | - | 1 | 72 | Chr. 12 - 80942857 - 80945374 [-] on Build GRCm38 | |

Gm20337
Genomic Map