siRNA Details
| Gene Symbol: 2210011K15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008708.1 | - | 2 | 914 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2210011K15Rik| Gene Symbol: 2210011K15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008708.1 | - | 2 | 914 | ||