siRNA Details
| Gene Symbol: 4933406C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016682.1 | - | 4 | 617 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716369.1 | - | 4 | 617 | Chr. 12 - 32926874 - 32953783 [-] on Build GRCm38 | |

4933406C10Rik
Genomic Map