siRNA Details
| Gene Symbol: Pus3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017552.1 | - | 1 | 1142 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY567142.1 | - | 1 | 185 | Chr. 9 - 35560442 - 35560865 [+] on Build GRCm38 | |

Pus3
Genomic Map