siRNA Details
| Gene Symbol: 6330571C24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018247.1 | - | 1 | 348 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY718710.1 | - | 1 | 348 | Chr. 11 - 77212406 - 77213206 [-] on Build GRCm38 | |

6330571C24Rik
Genomic Map