siRNA Details
| Gene Symbol: 9430012M22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020412.1 | - | 1 | 358 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY721588.1 | - | 1 | 358 | Chr. 3 - 55290944 - 55291595 [+] on Build GRCm38 | |

9430012M22Rik
Genomic Map