siRNA Details
Gene Symbol: Slc2a4rg-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045164.1 | 1 | 269 | |||
GenBank mRNA | AK082896.1 | - | 1 | 269 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc2a4rg-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045164.1 | 1 | 269 | |||
GenBank mRNA | AK082896.1 | - | 1 | 269 |