siRNA Details
| Gene Symbol: Slc2a4rg-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045164.1 | 1 | 536 | |||
| GenBank mRNA | AK082896.1 | - | 1 | 536 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc2a4rg-ps| Gene Symbol: Slc2a4rg-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045164.1 | 1 | 536 | |||
| GenBank mRNA | AK082896.1 | - | 1 | 536 | ||