siRNA Details
| Gene Symbol: 1700021P04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006227.1 | - | 1 | 120 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706313.1 | - | 1 | 120 | Chr. 19 - 24064793 - 24066452 [+] on Build GRCm38 | |

1700021P04Rik
Genomic Map