siRNA Details
| Gene Symbol: A530030E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079970.1 | - | 1 | 1195 | ||
| AK080099.1 | - | 1 | 1192 | |||
| BB220605.2 | - | 1 | 545 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A530030E21Rik| Gene Symbol: A530030E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079970.1 | - | 1 | 1195 | ||
| AK080099.1 | - | 1 | 1192 | |||
| BB220605.2 | - | 1 | 545 | |||