siRNA Details
| Gene Symbol: 2700046A07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037693.1 | 3 | 1945 | |||
| GenBank mRNA | AK012381.1 | - | 3 | 1874 | ||
| BC072581.1 | - | 3 | 1744 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2700046A07Rik| Gene Symbol: 2700046A07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037693.1 | 3 | 1945 | |||
| GenBank mRNA | AK012381.1 | - | 3 | 1874 | ||
| BC072581.1 | - | 3 | 1744 | |||