siRNA Details
| Gene Symbol: 2700046A07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037693.1 | 3 | 849 | |||
| GenBank mRNA | AK012381.1 | - | 3 | 778 | ||
| BC072581.1 | - | 3 | 648 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CD553290.1 | - | 3 | 448 | Chr. 18 - 62752831 - 62755946 [-] on Build GRCm38 | |

2700046A07Rik
Genomic Map