siRNA Details
| Gene Symbol: Gm20356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040450.1 | 2 | 204 | |||
| GenBank mRNA | AK139930.1 | - | 2 | 205 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY726125.1 | - | 2 | 205 | Chr. 3 - 74011032 - 74017277 [-] on Build GRCm38 | |

Gm20356
Genomic Map