siRNA Details
| Gene Symbol: Zfp866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133399.1 | - | 112 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB617032.1 | - | 2 | 108 | Chr. 8 - 69763368 - 69791175 [-] on Build GRCm38 | |

Zfp866
Genomic Map