siRNA Details
| Gene Symbol: 4921509O07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045501.1 | 4 | 482 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4921509O07Rik| Gene Symbol: 4921509O07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045501.1 | 4 | 482 | |||